missing translation for 'onlineSavingsMsg'
Learn More

Ethanediol, Extra Pure, SLR, Fisher Chemical™

Product Code. 10122000 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
Amber glass bottle
HDPE plastic bottle
Plastic container
Unit Size:
1 L
2.5 L
25 L
500mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10122000 1 L Amber glass bottle 1 L
10395700 500 mL HDPE plastic bottle 500mL
10274430 2.5 L Amber glass bottle 2.5 L
10346420 2.5 L HDPE plastic bottle 2.5 L
10000130 25 L Plastic container 25 L
10000130 25 L Plastic container 25 L
10000130 25 L Plastic container 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 10122000 Supplier Fisher Chemical Supplier No. E/0350/15
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

C2H6O2, CAS Number-107-21-1, ethylene alcohol, glycol alcohol, 2-hydroxyethanol, 1,2-ethanediol, glycol, fridex, monoethylene glycol, tescol, 1,2-dihydroxyethane, ethylene glycol, 2.5L, 196 deg.C, CHEBI:30742, Colorless, 62.06g/mol, LYCAIKOWRPUZTN-UHFFFAOYSA-N, ethane-1,2-diol, -13 deg.C, 2885 | CAS: 107-21-1 | C2H6O2 | 62.068 g/mol

C2H6O2, CAS Number-107-21-1, ethylene alcohol, glycol alcohol, 2-hydroxyethanol, 1,2-ethanediol, glycol, fridex, monoethylene glycol, tescol, 1,2-dihydroxyethane, ethylene glycol, 2.5L, 196 deg.C, CHEBI:30742, Colorless, 62.06g/mol, LYCAIKOWRPUZTN-UHFFFAOYSA-N, ethane-1,2-diol, -13 deg.C, 2885

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
MDL Number 2885
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Melting Point -13°C
Color Colorless
pH 5.5 to 7.5
Boiling Point 196°C
Packaging Amber glass bottle
Quantity 1 L
Vapor Density 2.14
Formula Weight 62.06g/mol
Vapor Pressure 0.12mmHg at 20°C
Physical Form Viscous Liquid
compliance-icons compliance-icons
Product Identifier
  • Ethylene glycol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.