missing translation for 'onlineSavingsMsg'
Learn More

Ethanethiol, 97%

Product Code. 15481087
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
Product Code. Quantity unitSize
15481087 25 mL 25mL
15491087 100 mL 100mL
15401097 500 mL 500mL
3 options
This item is not returnable. View return policy

Product Code. 15481087

Brand: Thermo Scientific Alfa Aesar 022585.AC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75-08-1 | C2H6S | 62.13 g/mol

Ethanethiol acts as a stabilizer for adhesives and an odorant for natural gas. It is used in the odorless liquefied petroleum gas (LPG) in order to find out the gas leaks. It is used as key starting material for the preparation of ethylsulfonic acid, diethyldisulfide and sodium ethylmercaptide. Further, it is also used in the underground mining industry as stench gas. In organic synthesis, it gives powerful nucleophile set- in the presence of sodium hydroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-08-1
Molecular Formula C2H6S
Molecular Weight (g/mol) 62.13
MDL Number MFCD00004887
InChI Key DNJIEGIFACGWOD-UHFFFAOYSA-N
Synonym ethyl mercaptan, mercaptoethane, thioethanol, ethyl hydrosulfide, ethyl sulfhydrate, ethyl thioalcohol, thioethyl alcohol, 1-mercaptoethane, aethanethiol, aethylmercaptan
PubChem CID 6343
IUPAC Name ethanethiol
SMILES CCS

Specifications

Melting Point -148°C
Color Colorless
Density 0.84 g/mL
Boiling Point 35°C
Flash Point -48°C (-55°F)
Odor Unpleasant
Linear Formula CH3CH2SH
Refractive Index 1.431
Quantity 25 mL
UN Number UN2363
Beilstein 773638
Merck Index 14,3726
Solubility Information Soluble in alcohol,ether,waterMiscible with alcohol,petroleum naphtha,acetone,dilute alkali and ether; Slightly miscible with water.
Formula Weight 62.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethanethiol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethanethiol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Flammable liquid Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contains : Ethanethiol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.