missing translation for 'onlineSavingsMsg'
Learn More

Ethanethiol, TRC

Product Code. 30388030
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
Product Code. Quantity unitSize
30388030 250 mL 250mL
1 options
This item is not returnable. View return policy

Product Code. 30388030

Brand: Toronto Research Chemicals TRCE676470250ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethanethiol
CAS 75-08-1
Purity Grade Notes GC
Molecular Formula C2 H6 S
Quantity 250 mL
Synonym Ethyl hydrosulfide, Ethyl mercaptan, Ethyl thioalcohol, Mercaptoethane, NSC 93877, Thioethanol, Thioethyl alcohol
InChI Formula InChI=1S/C2H6S/c1-2-3/h3H,2H2,1H3
SMILES CCS
IUPAC Name ethanethiol
Molecular Weight (g/mol) 62.134
Formula Weight 62.019
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.