missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, 96%, technical, denaturated with 5% methanol

Product Code. 13247983 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
Unit Size:
1 L
2.5 L
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13247983 1 L 1 L
13257983 2.5 L 2.5 L
13267983 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 13247983 Supplier Thermo Scientific Acros Supplier No. 444140010
  • ONLINE EXCLUSIVE PRICE
    32.25 €
    31.30 € / 1 L
    Save 0.95 €
    2% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

CAS: 64-17-5 | C2H6O | 46.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -117.0°C
Density 0.8000 g/mL
Quantity 1 L
Flash Point 12°C
Packaging Glass Bottle
Refractive Index 1.3610 to 1.3630
Assay Percent Range 96%
Specific Gravity 0.8
Linear Formula C2H5OH
Merck Index 14, 3761
Boiling Point 78.0°C
Solubility Information Solubility aq. soln.: completely soluble.
Formula Weight 46.06
Grade Technical
Chemical Name or Material Ethanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethanol, 96% denaturated with 5% methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H371-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P308-IF exposed or concerned:Call a POISON CENTERor doctor/ physician if you feel unwell.

For Manufacturing and Laboratory Use. Not for beverage use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.