missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, 99.8%, for Analysis, Absolute

Product Code. 11934051 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11934051 2.5 L 2.5 L
11924051 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 11934051 Supplier Thermo Scientific Acros Supplier No. 445730025
  • ONLINE EXCLUSIVE PRICE
    296.00 €
    273.00 € / 2.5 L
    Save 23.00 €
    7% Off
View all promotions
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

99.8% Ethanol for Analysis in a glass bottle. | CAS: 64-17-5 | C2H6O | 46.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • 99.8% Ethanol for Analysis, Absolute
  • GHS P Statement: Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower. Wear eye protection/face protection. Wash face, hands and any exposed skin thoroughly after handling. If eye irritation persists: Get medical advice/attention.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -114.0°C
Density 0.7900 g/mL
Quantity 2.5 L
Flash Point 12°C
Packaging Glass Bottle
Assay Percent Range 99.8%
Specific Gravity 0.79
Linear Formula CH3CH2OH
Viscosity 1.2 mPa.s (20°C)
Merck Index 15, 3814
Boiling Point 78.0°C
Solubility Information Solubility aq. soln.: completely soluble.
Formula Weight 46.07
Grade Analytical
Chemical Name or Material Ethanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

For Manufacturing and Laboratory Use. Not for beverage use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.