missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol 300mg/100mL in Water, LoGiCal
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Ethanol 300mg/100mL in Water
Specifications
Specifications
| Chemical Name or Material | Ethanol |
| CAS | 64-17-5 |
| Quantity | 1 mL |
| Molecular Formula | C2 H6 O |
| Formulation | Single Solution |
| Synonym | Ethanol, Dehydrated Alcohol, Ethanol (9 CI, ACI), Ethyl alcohol (6 CI, 7 CI, 8 CI), 100 C.NPA, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A |
| InChI Formula | InChI=1 S/C2H6O/c1-2-3/h3H,2H2,1H3 |
| SMILES | CCO |
| IUPAC Name | ethanol |
| Molecular Weight (g/mol) | 46.07 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur