missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, 95+%, pure, denat. with 5% wood spirit

Product Code. 10329323 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Unit Size:
1 L
10 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10329323 10 L 10 L
10692033 1 L 1 L
10404985 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 10329323 Supplier Thermo Scientific Acros Supplier No. 365780100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

CAS: 64-17-5 | C2H6O | 46.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5 , 67-56-1
Molecular Formula C2H6O
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym grain alcohol, algrain, ethyl hydrate, ethyl hydroxide, methylcarbinol, ethyl alcohol, alcohol, tecsol, anhydrol, alcohol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -117.0°C
Density 0.8000 g/mL
Quantity 10 L
Flash Point 12°C
Packaging Plastic Drum
Refractive Index 1.3610 to 1.3630
Specific Gravity 0.8
Linear Formula C2H5OH
Boiling Point 78.0°C
Solubility Information Solubility aq. soln.: miscible. Other solubilities: miscible with many organic solvents
Molecular Weight (g/mol) 46.07
Formula Weight 46.06
Grade Pure
Chemical Name or Material Ethanol
Show More Show Less

For Manufacturing and Laboratory Use. Not for beverage use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.