missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Melting Point | -114°C |
| Color | Colorless |
| Boiling Point | 78°C |
| Quantity | 2.5 L |
| Packaging | Amber Glass Bottle |
| Physical Form | Liquid |
| Molecular Weight (g/mol) | 46.069 |
| Formula Weight | 46.07g/mol |
Safety and Handling
Product Identifier
- Ethanol (95% v/v)
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?