missing translation for 'onlineSavingsMsg'
Learn More

Ethanol 95% (v/v), Extra Pure, SLR, Fisher Chemical™

Product Code. 10572143 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
10 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Plastic Container
Plastic-coated Glass Bottle
Unit Size:
1 L
10 L
2.5 L
5 L
500mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10572143 2.5 L HDPE Bottle 2.5 L
10510153 500 mL Amber Glass Bottle 500mL
10681533 1 L Amber Glass Bottle 1 L
10040602 2.5 L Amber Glass Bottle 2.5 L
12666697 2.5 L Plastic-coated Glass Bottle 2.5 L
10305752 5 L HDPE Bottle 5 L
10447492 10 L Plastic Container 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 10572143 Supplier Fisher Chemical Supplier No. E/0500DF/C17
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
MDL Number 3568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Spezifikation

Melting Point -114°C
Color Colorless
Quantity 2.5 L
Packaging HDPE Bottle
Physical Form Liquid
Boiling Point 78°C
Molecular Weight (g/mol) 46.07
Formula Weight 46.07g/mol
compliance-icons compliance-icons
Produktidentifikator
  • Ethanol (95% v/v)
Signalwort
  • Danger
Gefahrenkategorie
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Gefahrenhinweis
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.