missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, 96%, technical, not denaturated

Product Code. 13217993 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 L
Packaging:
Plastic Drum
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13217993 5 L Plastic Drum 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 13217993 Supplier Thermo Scientific Acros Supplier No. 444190050
  • ONLINE EXCLUSIVE PRICE
    142.00 €
    132.00 € / 5 L
    Save 10.00 €
    7% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

CAS: 64-17-5 | C2H6O | 46.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Spezifikation

Melting Point -114.0°C
CAS 64-17-5
Density 0.8000 g/mL
Quantity 5 L
Flash Point 17°C
Packaging Plastic Drum
Assay Percent Range 96%
Specific Gravity 0.8
Molecular Formula C2H6O
Linear Formula C2H5OH
Viscosity 1.2 mPa.s (20°C)
Merck Index 15, 3814
Boiling Point 78.0°C
Solubility Information Solubility aq. soln.: soluble.
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.07
Formula Weight 46.07
Grade Technical
Chemical Name or Material Ethanol
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Produktidentifikator
  • Ethanol 96%, technical, duty free
Signalwort
  • Danger
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Gefahrenhinweis
  • H319-Causes serious eye irritation.
  • H225-Highly flammable liquid and vapour.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

For Manufacturing and Laboratory Use. Not for beverage use

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.