missing translation for 'onlineSavingsMsg'
Learn More

Ethanol 96% v/v, Certified AR for analysis, meets analytical specification of BP and Ph. Eur., Fisher Chemical™

Product Code. 10703753 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Metal Can
Unit Size:
2.5 L
25 L
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10703753 5 L HDPE Bottle 5 L
10332562 2.5 L Amber Glass Bottle 2.5 L
10608932 25 L Metal Can 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 10703753 Supplier Fisher Chemical Supplier No. E/0555DF/21
  • ONLINE EXCLUSIVE PRICE
    206.00 €
    194.00 € / 5 L
    Save 12.00 €
    5% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

C2H6O, CAS Number-64-17-5, 2.5L, 78 deg.C, CHEBI:16236, Colorless, 46.07g/mol, LFQSCWFLJHTTHZ-UHFFFAOYSA-N, -114 deg.C, 3568, Amber glass bottle, Liquid, 702

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
MDL Number 3568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -114°C
Color Colorless
Quantity 5 L
Packaging HDPE Bottle
Physical Form Liquid
Boiling Point 78°C
Formula Weight 46.07g/mol
compliance-icons compliance-icons
Product Identifier
  • Ethanol (96% v/v)
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contain: Ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.