missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, Absolute (200 Proof), Molecular Biology Grade, Fisher BioReagents™

Product Code. 16695992 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 L
Unit Size:
0.1 L
2.50 L
4 L
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16695992 4 L 4 L
16685992 100 mL 0.1 L
16606002 500 mL 500mL
10644795 500 mL 500mL
16688242 2.5 L 2.50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 16695992 Supplier Fisher BioReagents Supplier No. BP28184
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Meets ACS Specifications

  • Meets ACS Specifications
  • For nucleic acid purification and precipitation
  • Ultrapure molecular biology grade ethanol is used for the purification and precipitation of biomolecules such as nucleic acids and proteins
  • It is also used in histology to prepare staining and destaining reagents and for dehydrating tissues prior to embedding
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -114°C
Color Colorless
Quantity 4 L
Flash Point 12°C (53.6°F)
Identification Pass Test
Infrared Spectrum Conforms to reference
Physical Form Liquid
Residue after Evaporation 0.001% max.
Assay Percent Range Assay: ≥99.5% (v/v)
Titratable Acid 0.0005 mEq /g max.
Titratable Base 0.0002 mEq /g max.
Linear Formula C2H5OH
Water 0.2% max.
Merck Index 15, 3814
Boiling Point 78°C
Molecular Weight (g/mol) 46.069
Formula Weight 46.07
Percent Purity ≥99.5%
Grade DNase-, RNase- and Protease-Free
Chemical Name or Material Ethanol, Absolute (200 Proof)
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Molecular Biology Grade Ethanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.