missing translation for 'onlineSavingsMsg'
Learn More

Ethanol Absolute, 99.9%, BAKER ANALYZED™, J.T. Baker™

Product Code. 15578454 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
HDPE Bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15578454 1 L HDPE Bottle 1 L
13268633 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 15578454 Supplier JT Baker Supplier No. 8025.1000PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Exceeds ACS specifications to provide improved consistency and characterization of the reagent. J.T. Baker™ Ethanol Absolute, 99.9%, BAKER ANALYZED™ meets EPA method 130.2 for testing of water hardness and meets US FDA requirements for use in the pharmaceutical industry.

BAKER ANALYZED™ reagents are a grade of high quality chemicals for laboratory and specialized industrial use. Every BAKER ANALYZED™ Reagent label provides an Actual Lot Analysis as a guarantee of purity and consistency. Analytical methods used for these determinations are based on standard test procedures described in technical publications.

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07

Specifications

Melting Point −114.5°C
Color Colorless
Density 0.79g/cm3 at 20°C
Quantity 1 L
Explosion Limit Lower: 3.5 Vol %, Upper: 15 Vol %
Flash Point 12°C
Ignition Point 425°C
Odor Alcohol
Packaging HDPE Bottle
Physical Form Liquid
Residue after Evaporation Max. 0.001%
Titratable Acid Max. 0.0005 meq/g
Titratable Base Max. 0.0002 meq/g
Viscosity 1.2 mPas at 20°C
Water Max. 0.2%
Boiling Point 78°C
Solubility Information 1g/L in water at 20°C
Percent Purity Min. 99.9% v/v
Chemical Name or Material Ethanol, Absolute
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethanol Absolute
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.