missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- UV-absorbance (1cm vs hexane)
- at 220 nm max. 0.30
- at 230 nm max. 0.18
- at 240 nm max. 0.08
- at 270 nm max. 0.02
- 220-270 nm (no maxima) Passes Test
Specifications
Specifications
| Melting Point | -114°C |
| CAS | 64-17-5 |
| Appearance | Passes Test |
| Density | 0.804 - 0.809 g/mL (at 20°C) |
| Quantity | 2.5 L |
| Identification | Passes Test |
| Packaging | Plastic Bottle |
| Residue after Evaporation | max. 0.0015% |
| Assay Percent Range | 96.0 to 97.2% (v/v) Passes Test |
| Titratable Acid | Passes Test |
| Show More |
Safety and Handling
Product Identifier
- Ethanol min. 95% by volume
- Danger
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P403+P235-Store in a well-ventilated place. Keep cool.
- MIXTURE LIST-Contain: Ethanol Absolute
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?