missing translation for 'onlineSavingsMsg'
Learn More

Ethanol (Untaxed), Puriss., meets analytical spec. of BP, Ph. Eur., 96% (v/v), for license holders only, Solstice
BIOPHARMA

Product Code. 15662490
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
30 kg
4 x 2.5 L
4 x 5 L
160 kg
Packaging:
Glass Bottle
Metal Drum
Plastic Bottle
Poly Bottle
Stainless Steel Can
Unit Size:
1 L
160kg
2.5 L
30kg
5 L
Pack of 4
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15662490 4 x 5 L Poly Bottle Pack of 4
15622490 1 L Glass Bottle 1 L
15612490 1 L Poly Bottle 1 L
15632490 2.5 L Plastic Bottle 2.5 L
15672490 5 L Poly Bottle 5 L
15652490 4 x 2.5 L Plastic Bottle Pack of 4
15642490 30 kg Stainless Steel Can 30kg
15602490 160 kg Metal Drum 160kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 15662490 Supplier Honeywell-Riedel-de Haen Supplier No. 241064X5L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -114°C (literature)
CAS Max % 100.0000%
Density 0.789 g/mL at 25 °C(lit.)
Quantity 4 x 5 L
Flash Point 12°C
Packaging Poly Bottle
Linear Formula NaHSO3
MDL Number MFCD00003530
UN Number UN1170
Beilstein 1718733
Boiling Point 78°C to 79°C
Formula Weight 46
Percent Purity 96%
Grade puriss.
Chemical Name or Material Ethanol (Untaxed), Puriss., meets analytical specification of BP, Ph. Eur., 96% (v/v), for license holders only
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethanol puriss., meets analytical specification of Ph. Eur., BP, 96% (v/v)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.