missing translation for 'onlineSavingsMsg'
Learn More

Ethanol (Untaxed), Puriss. p.a., ACS Reagent, Reag. Ph. Eur., 96% (v/v), for license holders only, Honeywell Riedel-de Haën™

Product Code. 15643770
Change view
Click to view available options
Quantity:
1 L
5 L
30 kg
4 x 2.5 L
4 x 5 L
6 x 1 L
Packaging:
Poly Bottle
Stainless Steel Can
Unit Size:
1 L
30kg
5 L
Pack of 4
Pack of 6
Product Code. Quantity Packaging unitSize
15643770 4 x 2.5 L Poly Bottle Pack of 4
15603770 1 L Poly Bottle 1 L
15663770 5 L Poly Bottle 5 L
15673770 6 x 1 L Poly Bottle Pack of 6
15653770 4 x 5 L Poly Bottle Pack of 4
15633770 30 kg Stainless Steel Can 30kg
6 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Product Code. 15643770

Brand: Honeywell-Riedel-de Haen 322944X2.5L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00000881
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -114°C (literature)
CAS Max % 100.0000%
Density 0.789 g/mL at 25 °C(lit.)
Boiling Point 78°C to 79°C
Quantity 4 x 2.5 L
Flash Point 12°C
Packaging Poly Bottle
Linear Formula CH2Cl2
UN Number UN1170
Formula Weight 46
Percent Purity 96%
Grade ACS Reagent/Reagent Ph. Eur.
Chemical Name or Material Ethanol (Untaxed), Puriss. p.a., ACS Reagent, Reag. Ph. Eur., 96% (v/v), for license holders only
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • ethanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P264-Wash skin thoroughly after handling.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P242-Use only non-sparking tools.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P233-Keep container tightly closed.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P240-Ground/bond container and receiving equipment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.