missing translation for 'onlineSavingsMsg'
Learn More

Ethanolamine, 99%, AcroSeal™

Product Code. 10473264 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10473264 800 mL AcroSeal™ Glass bottle 800mL
10754775 100 mL AcroSeal™ Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10473264 Supplier Thermo Scientific Acros Supplier No. 427258000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 141-43-5 | C2H7NO | 61.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-43-5
Molecular Formula C2H7NO
Molecular Weight (g/mol) 61.08
MDL Number MFCD00008183
InChI Key HZAXFHJVJLSVMW-UHFFFAOYSA-N
Synonym ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine
PubChem CID 700
ChEBI CHEBI:16000
SMILES NCCO

Specifications

Melting Point 10.3°C
Density 1.0120g/mL
Boiling Point 170.0°C
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging AcroSeal™ Glass bottle
Linear Formula H2NCH2CH2OH
Refractive Index 1.4530 to 1.4550
Quantity 800 mL
Merck Index 15, 3782
Specific Gravity 1.012
Solubility Information Solubility in water: soluble. Other solubilities: miscible with methanol and acetone, soluble in benzene, ether, ccl4 and n-heptane
IUPAC Name 2-aminoethanol
Viscosity 24 mPa.s (20°C)
Formula Weight 61.08
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethanolamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethanolamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.