missing translation for 'onlineSavingsMsg'
Learn More

Ethanolamine, ACS, 99+%

Product Code. 11465160
Change view
Click to view available options
Quantity:
500 mL
4 x 500 mL
Unit Size:
2000mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465160 4 x 500 mL 2000mL
11455160 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11465160 Supplier Thermo Scientific Alfa Aesar Supplier No. 036260.AY
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 141-43-5 | C2H7NO | 61.08 g/mol

It is also used in messenger molecules such as palmitoylethanolamide, which have an effect on CB1 receptors. Ethanolamine is used as a scrubber for the removal of methyl bromide. Polyaniline doping ethanolamine is used for modified electrode preparation to measure uric acid in human body with the presence of antiscorbutic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-43-5
Molecular Formula C2H7NO
Molecular Weight (g/mol) 61.08
MDL Number MFCD00008183
InChI Key HZAXFHJVJLSVMW-UHFFFAOYSA-N
Synonym ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine
PubChem CID 700
ChEBI CHEBI:16000
SMILES NCCO

Specifications

Melting Point 10°C
Density 1.012 g/mL
Boiling Point 169°C to 170°C
Flash Point 93°C (199°F)
Assay Percent Range >99+%
Odor Ammonia-like
Refractive Index 1.454
Quantity 4 x 500 mL
UN Number UN2491
Beilstein 505944
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,3727
Solubility Information Miscible in water.
IUPAC Name 2-aminoethanol
Formula Weight 61.08
Percent Purity ≥99%
Grade ACS
Physical Form Liquid
Chemical Name or Material Ethanolamine, For analysis ACS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.