missing translation for 'onlineSavingsMsg'
Learn More

Ethanolamine hydrochloride, 99+%

Product Code. 10761145 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10761145 1 kg Glass bottle 1kg
10226750 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10761145 Supplier Thermo Scientific Acros Supplier No. 217040010
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2002-24-6 | C2H7NO·HCl | 97.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2002-24-6
Molecular Formula C2H7NO·HCl
Molecular Weight (g/mol) 97.55
MDL Number MFCD00012892
InChI Key PMUNIMVZCACZBB-UHFFFAOYSA-N
Synonym 2-aminoethanol hydrochloride, ethanolamine hydrochloride, ethanolamine hcl, mea hydrochloride, monoethanolamine hydrochloride, ethanolamine chloride, colamine hydrochloride, ethanol, 2-amino-, hydrochloride, ethanolamine-hcl, 2-hydroxyethylammonium chloride
PubChem CID 74819
IUPAC Name 2-aminoethanol;hydrochloride
SMILES C(CO)N.Cl

Specifications

Melting Point 84°C to 86°C
Color Beige or White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (Argentometry)
Packaging Glass bottle
Linear Formula H2NCH2CH2OH·HCl
Quantity 1 kg
Beilstein 04, 275
Merck Index 15, 3782
Solubility Information Solubility in water: soluble
Formula Weight 97.55
Percent Purity 99+%
Physical Form Agglomerating Crystals
Chemical Name or Material Ethanolamine hydrochloride, p.a.
Show More Show Less
compliance-icons
Product Identifier
  • Ethanolamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.