missing translation for 'onlineSavingsMsg'
Learn More

Ethanolamine, TRC

Product Code. 30369547
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30369547 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30369547 Supplier Toronto Research Chemicals Supplier No. TRCE67650025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethanolamine
CAS 141-43-5
Purity Grade Notes HPLC
Molecular Formula C2 H7 N O
Quantity 25 g
Synonym 2-Aminoethanol, 1-Amino-2-hydroxyethane, 2-Amino-1-ethanol, 2-Aminoethyl alcohol, 2-Ethanolamine, 2-Hydroxyethanamine, 2-Hydroxyethylamine, Aminoethanol, Colamine, ETA, Envision Conditioner PDD 9020, Ethanolamine, Ethylolamine, Glycinol, Kolamin, MEA, MEA (alcohol), MEA 90, MEA-LCI, Mealan, Monoethanolamine, Olamine, R 100, R 100 (amine), Seramine, β-Aminoethanol, β-Aminoethyl alcohol, β-Ethanolamine, β-Hydroxyethylamine
InChI Formula InChI=1S/C2H7NO/c3-1-2-4/h4H,1-3H2
SMILES NCCO
IUPAC Name 2-aminoethanol
Molecular Weight (g/mol) 61.08
Formula Weight 61.0528
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.