missing translation for 'onlineSavingsMsg'
Learn More

Ethaverine, TRC

Product Code. 17903465
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17903465 100 mg 100mg
17962455 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17903465 Supplier Toronto Research Chemicals Supplier No. TRCE676700100MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 486-47-5
Molecular Formula C24 H29 N O4
Molecular Weight (g/mol) 395.49
Synonym 1-(3,4-Diethoxybenzyl)-6,7-diethoxyisoquinoline, Perparine, 1-(3,4-Diethoxybenzyl)-6,7-diethoxyisoquinoline, 6,7-Diethoxy-1-(3,4-diethoxybenzyl)isoquinoline, Barbonin, Barbonine, Dyscural, Ethylpapaverine, Ethylpapaverine
IUPAC Name 1-[(3,4-diethoxyphenyl)methyl]-6,7-diethoxyisoquinoline
SMILES CCOc1ccc(Cc2nccc3cc(OCC)c(OCC)cc23)cc1OCC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C24H29NO4/c1-5-26-21-10-9-17(14-22(21)27-6-2)13-20-19-16-24(29-8-4)23(28-7-3)15-18(19)11-12-25-20/h9-12,14-16H,5-8,13H2,1-4H3
Formula Weight 395.2097
Percent Purity >95%
Chemical Name or Material Ethaverine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.