missing translation for 'onlineSavingsMsg'
Learn More

Ethidium bromide, 95%, pure

Product Code. 10742791 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10742791 1 g Glass bottle 1g
10042120 5 g Glass bottle 5g
11976611 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10742791 Supplier Thermo Scientific Acros Supplier No. 170960010
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 1239-45-8 | C21H20BrN3 | 394.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1239-45-8
Molecular Formula C21H20BrN3
Molecular Weight (g/mol) 394.32
MDL Number MFCD00011724
InChI Key ZMMJGEGLRURXTF-UHFFFAOYSA-N
Synonym ethidium bromide, homidium bromide, dromilac, 3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide, ethydium bromide, 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide, etbr, 2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide, 2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide, unii-059nuo2z1l
PubChem CID 14710
ChEBI CHEBI:4883
IUPAC Name 5-ethyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide
SMILES [Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12

Specifications

Melting Point 260.0°C to 262.0°C
Color Purple to Red
pH ∼4.5 (1% aq. soln. at 20°C)
Flash Point >100°C
Loss on Drying 5.0% max.
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Argentometry)
Packaging Glass bottle
Quantity 1 g
Merck Index 15,4769
Solubility Information (1% in water) Clear red
Formula Weight 394.3
Percent Purity 95%
Grade Pure
Physical Form Crystalline Powder
Chemical Name or Material Ethidium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.