Learn More
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
| Melting Point | 260.0°C to 262.0°C |
| Color | Purple to Red |
| pH | ∼4.5 (1% aq. soln. at 20°C) |
| Flash Point | >100°C |
| Loss on Drying | 5.0% max. |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 94% min. (Argentometry) |
| Packaging | Glass bottle |
| Quantity | 25 g |
| Merck Index | 15,4769 |
| Mostrar más |
Seguridad y manipulación
- Ethidium bromide
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 4
- Germ cell mutagenicity Category 2
- H302-Harmful if swallowed.
- H330-Fatal if inhaled.
- H341-Suspected of causing genetic defects.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: 3,8-Diamino-1-ethyl-6-phenylphenantridinium bromide
RUO – Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.