missing translation for 'onlineSavingsMsg'
Learn More

Ethidium bromide, 99.11%, MP Biomedicals™

Product Code. 11491631 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Unit Size:
10g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491631 1 g 1g
11401641 5 g 5g
11411641 10 g 10g
11421641 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11491631 Supplier MP Biomedicals Supplier No. 0219020201
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1239-45-8
Molecular Formula C21H20BrN3
Molecular Weight (g/mol) 394.32
MDL Number MFCD00011724
InChI Key ZMMJGEGLRURXTF-UHFFFAOYSA-N
Synonym ethidium bromide, homidium bromide, dromilac, 3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide, ethydium bromide, 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide, etbr, 2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide, 2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide, unii-059nuo2z1l
PubChem CID 14710
ChEBI CHEBI:4883
IUPAC Name 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
SMILES [Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12

Specifications

Color Purple-Red
Assay Percent Range 1239-45-8
Quantity 1 g
Formula Weight 394.3
Chemical Name or Material Ethidium bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.