missing translation for 'onlineSavingsMsg'
Learn More

Ethionamide, TRC

Product Code. 30384427
Change view
Click to view available options
Quantity:
50 mg
5 g
25 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384427 5 g 5g
30376393 50 mg -
30372883 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384427 Supplier Toronto Research Chemicals Supplier No. TRCE8904205G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 536-33-4
Molecular Formula C8 H10 N2 S
Molecular Weight (g/mol) 166.24
Synonym Ethionamide
IUPAC Name 2-ethylpyridine-4-carbothioamide
SMILES CCc1cc(ccn1)C(=S)N

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H10N2S/c1-2-7-5-6(8(9)11)3-4-10-7/h3-5H,2H2,1H3,(H2,9,11)
Formula Weight 166.0565
Chemical Name or Material Ethionamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.