missing translation for 'onlineSavingsMsg'
Learn More

Ethiprole, TRC

Product Code. 30375272
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30375272 5 g
30372473 1 g
30384963 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30375272 Supplier Toronto Research Chemicals Supplier No. TRCE6783205G
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 181587-01-9
Molecular Formula C13 H9 Cl2 F3 N4 O S
Molecular Weight (g/mol) 397.2
Synonym 5-Amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-(ethylsulfinyl)-1H-pyrazole-3-carbonitrile, 5-Amino-1-[2,6-Dichloro-4-(trifluoromethyl)phenyl]-4-(ethylsulfinyl)pyrazole-3-carbonitrile, Ethiprole, Kirappu, RPA 107382
IUPAC Name 5-amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-ethylsulfinylpyrazole-3-carbonitrile
SMILES CCS(=O)c1c(N)n(nc1C#N)c2c(Cl)cc(cc2Cl)C(F)(F)F

Specifications

Quantity 5 g
InChI Formula InChI=1S/C13H9Cl2F3N4OS/c1-2-24(23)11-9(5-19)21-22(12(11)20)10-7(14)3-6(4-8(10)15)13(16,17)18/h3-4H,2,20H2,1H3
Formula Weight 395.9826
Chemical Name or Material Ethiprole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.