missing translation for 'onlineSavingsMsg'
Learn More

Ethopropazine hydrochloride, TRC

Product Code. 30382848
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382848 250 mg
30373104 25 mg
30369377 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382848 Supplier Toronto Research Chemicals Supplier No. TRCE677660250MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1094-08-2
Molecular Formula C19 H24 N2 S . Cl H
Molecular Weight (g/mol) 348.93
Synonym 10H-Phenothiazine-10-ethanamine, N,N-diethyl-α-methyl-, hydrochloride (1:1), 10H-Phenothiazine-10-ethanamine, N,N-diethyl-α-methyl-, monohydrochloride (9CI), Phenothiazine, 10-[2-(diethylamino)propyl]-, hydrochloride (7CI), Phenothiazine, 10-[2-(diethylamino)propyl]-, monohydrochloride (8CI), Dibutil, Ethopropazine hydrochloride, Isothazine hydrochloride, Lysivane, Lysivane hydrochloride, NSC 169467, NSC 64074, Pardisol, Parfezin, Parphezein, Parphezin, Parsidol, Parsidol hydrochloride, Parsidol monohydrochloride, Parsitan, Parsotil, Profenamine hydrochloride, Profenamine monohydrochloride, Rodipal
IUPAC Name N,N-diethyl-1-phenothiazin-10-ylpropan-2-amine;hydrochloride
SMILES Cl.CCN(CC)C(C)CN1c2ccccc2Sc3ccccc13

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C19H24N2S.ClH/c1-4-20(5-2)15(3)14-21-16-10-6-8-12-18(16)22-19-13-9-7-11-17(19)21;/h6-13,15H,4-5,14H2,1-3H3;1H
Formula Weight 348.1427
Chemical Name or Material Profenamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.