missing translation for 'onlineSavingsMsg'
Learn More

Ethoxyamine hydrochloride, 98%

Product Code. 10740482 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10740482 10 g Glass bottle 10g
10615681 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10740482 Supplier Thermo Scientific Acros Supplier No. 409610100
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 3332-29-4 | C2H7NO·HCl | 97.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3332-29-4
Molecular Formula C2H7NO·HCl
Molecular Weight (g/mol) 97.54
MDL Number MFCD00012956
InChI Key NUXCOKIYARRTDC-UHFFFAOYSA-N
Synonym ethoxyamine hydrochloride, o-ethylhydroxylamine hydrochloride, hydroxylamine, o-ethyl-, hydrochloride, ethoxyamine hcl, hydroxylamine, o-ethyl-, hydrochloride 1:1, ethoxyaminehydrochloride, aminooxy ethane hydrochloride 1:1, etonh2.hcl, ethoxylamine hydrochloride, nh2och2ch3.hcl
PubChem CID 76850
IUPAC Name O-ethylhydroxylamine;hydrochloride
SMILES CCON.Cl

Specifications

Melting Point 130°C to 133°C
Color Gray
Infrared Spectrum Authentic
Assay Percent Range 99% min. (Argentometry)
Packaging Glass bottle
Linear Formula C2H5ONH2·HCl
Quantity 10 g
Solubility Information Solubility in water: soluble
Formula Weight 97.54
Percent Purity 99+%
Physical Form Crystals, Powder or Flakes
Chemical Name or Material Ethoxyamine hydrochloride, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethoxyamine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.