missing translation for 'onlineSavingsMsg'
Learn More

Ethoxybenzene, TRC

Product Code. 30387308
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30387308 25 mL 25mL
30383693 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30387308 Supplier Toronto Research Chemicals Supplier No. TRCE89064525ML
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-73-1
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.16
Synonym Phenetole, Ethoxybenzene, Ethyl Phenyl Ether, NSC 406706, Phenetol, Phenoxyethane, Phenyl Ethyl Ether
SMILES CCOc1ccccc1

Specifications

Quantity 25 mL
InChI Formula InChI=1S/C8H10O/c1-2-9-8-6-4-3-5-7-8/h3-7H,2H2,1H3
Formula Weight 122.0732
Chemical Name or Material Ethoxybenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.