missing translation for 'onlineSavingsMsg'
Learn More

Ethoxycarbonyl isothiocyanate, 95%

Product Code. 10757358 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10757358 25 g Glass bottle 25g
10208097 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Código de producto. 10757358 Proveedor Thermo Scientific Acros N.º de proveedor 187850250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 16182-04-0 | C4H5NO2S | 131.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 16182-04-0
Molecular Formula C4H5NO2S
Molecular Weight (g/mol) 131.15
MDL Number MFCD00004814
InChI Key BDTDECDAHYOJRO-UHFFFAOYSA-N
Synonym ethoxycarbonyl isothiocyanate, ethyl isothiocyanatoformate, carbethoxy isothiocyanate, ethoxycarbonylisothiocyanate, ethyl n-carbothioylcarbamate, ethyl n-sulfanylidenemethylidene carbamate, ethyl isothiocyanatidocarbonate, carbon isothiocyanatidic acid, ethyl ester, o-ethyl carbonisothiocyanatidate
PubChem CID 85320
IUPAC Name ethyl N-(sulfanylidenemethylidene)carbamate
SMILES CCOC(=O)N=C=S

Especificaciones

Density 1.1600g/mL
Boiling Point 56.0°C (18.0 mmHg)
Flash Point 50°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula SCNCOOCH2CH3
Quantity 25 g
Beilstein 03, III, 279
Fieser 04,223; 06,250; 07,146
Specific Gravity 1.16
Formula Weight 131.15
Percent Purity 95%
Chemical Name or Material Ethoxycarbonyl isothiocyanate
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.