missing translation for 'onlineSavingsMsg'
Learn More

Ethoxymethylene Malononitrile, TRC

Product Code. 30378935
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30378935 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30378935 Supplier Toronto Research Chemicals Supplier No. TRCE89245010G
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethoxymethylene Malononitrile
CAS 123-06-8
Molecular Formula C6 H6 N2 O
Quantity 10 g
Synonym Propanedinitrile, 2-(ethoxymethylene)-, Malononitrile, (ethoxymethylene)- (6CI,7CI,8CI), Propanedinitrile, (ethoxymethylene)- (9CI), 2-(Ethoxymethylene)propanedinitrile, (Ethoxymethylene)malonodinitrile, (Ethoxymethylene)malononitrile, (Ethoxymethylene)propanedinitrile, (Ethoxymethylidene)malononitrile, (β-Ethoxymethylene)malononitrile, 1,1-Dicyano-2-ethoxyethene, 1-Ethoxy-2,2-dicyanoethene, 1-Ethoxy-2,2-dicyanoethylene, 1-Ethoxymethylenemalonitrile, 2-(Ethoxymethylene)malononitrile, 2-Cyano-3-ethoxyacrylonitrile, 2-Ethoxy-1,1-ethenedicarbonitrile, Ethoxymethylenemalonitrile, NSC 27792, α-Cyano-β-ethoxyacrylonitrile
InChI Formula InChI=1S/C6H6N2O/c1-2-9-5-6(3-7)4-8/h5H,2H2,1H3
SMILES CCOC=C(C#N)C#N
IUPAC Name 2-(ethoxymethylidene)propanedinitrile
Molecular Weight (g/mol) 122.12
Formula Weight 122.048
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.