missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-aminocyclopropanecarboxylate hydrochloride, 98%

Product Code. 10793922 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10793922 25 g Glass bottle 25g
10407028 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10793922 Supplier Thermo Scientific Acros Supplier No. 432510250
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 42303-42-4 | C6H11NO2·ClH | 165.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42303-42-4
Molecular Formula C6H11NO2·ClH
Molecular Weight (g/mol) 165.62
MDL Number MFCD00190747
InChI Key XFNUTZWASODOQK-UHFFFAOYSA-N
Synonym ethyl 1-aminocyclopropanecarboxylate hydrochloride, 1-aminocyclopropane-1-carboxylic acid ethyl ester hydrochloride, h-acpc-oet.hcl, ethyl 1-aminocyclopropane-1-carboxylate hydrochloride, 1-amino-cyclopropyl-1-carboxylic acid ethyl ester hydrochloride, h-acpc-oet hcl, 1-aminocyclopropane-1-carboxylic acid ethyl ester, acpc-oet-hcl, h-acpc-oet?cl, pubchem14733
PubChem CID 386203
IUPAC Name ethyl 1-aminocyclopropane-1-carboxylate;hydrochloride
SMILES CCOC(=O)C1(CC1)N.Cl

Specifications

Melting Point 113.0°C to 114.0°C
Color White
Elemental Analysis 42.41 to 45.48% C, 8.24 to 8.66% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Formula Weight 165.62
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 1-aminocyclopropanecarboxylate hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 1-aminocyclopropanecarboxylate hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.