missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-Boc-DL-nipecotate, 97%, Thermo Scientific Chemicals

Product Code. 15475457
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15475457 5 g 5g
15465457 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15475457 Supplier Thermo Scientific Alfa Aesar Supplier No. H32704.06
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 130250-54-3 | Molecular Formula: C13H23NO4 | Molecular Weight: 257.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130250-54-3
Molecular Formula C13H23NO4
Molecular Weight (g/mol) 257.33
MDL Number MFCD04116274
InChI Key YCXCRFGBFZTUSU-UHFFFAOYSA-N
Synonym ethyl 1-boc-3-piperidinecarboxylate, 1-tert-butyl 3-ethyl piperidine-1,3-dicarboxylate, ethyl n-boc-piperidine-3-carboxylate, ethyl n-boc-3-piperidinecarboxylate, ethyl1-boc-3-piperidinecarboxylate, 1-boc-piperidine-3-carboxylic acid ethyl ester, ethyl-n-boc-piperidine-3-carboxylate, 1-tert-butyl 3-ethyl tetrahydro-1,3 2h-pyridinedicarboxylate, 1,3-piperidinedicarboxylic acid, 1-1,1-dimethylethyl 3-ethyl ester, piperidine-1,3-dicarboxylic acid 1-tert-butyl 3-ethyl ester
PubChem CID 357727
IUPAC Name 1-O-tert-butyl 3-O-ethyl piperidine-1,3-dicarboxylate
SMILES CCOC(=O)C1CCCN(C1)C(=O)OC(C)(C)C

Specifications

Melting Point 31°C to 35°C
Boiling Point 95°C (0.5 mmHg)
Quantity 5 g
Sensitivity Light and moisture sensitive
Formula Weight 257.33
Percent Purity 97%
Chemical Name or Material Ethyl 1-Boc-DL-nipecotate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.