missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-hydroxycyclopropanecarboxylate, 90%

Product Code. 10662572 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10662572 1 g Glass bottle 1g
11915081 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10662572 Supplier Thermo Scientific Acros Supplier No. 335300010
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 137682-89-4 | C6H10O3 | 130.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137682-89-4
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD01863257
InChI Key DPMGQZGXWRHYPE-UHFFFAOYSA-N
Synonym ethyl 1-hydroxycyclopropanecarboxylate, 1-hydroxycyclopropanecarboxylic acid ethyl ester, cyclopropanecarboxylic acid, 1-hydroxy-, ethyl ester, 1-ethoxycarbonyl cyclopropan-1-ol, ethyl-1-hydroxycyclopropane carboxylate, 1-hydroxy-cyclopropanecarboxylic acid ethyl ester
PubChem CID 2733706
IUPAC Name ethyl 1-hydroxycyclopropane-1-carboxylate
SMILES CCOC(=O)C1(O)CC1

Specifications

Color Yellow to Brown
Boiling Point 78.0°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula C3H4(OH)COOCH2CH3
Refractive Index 1.4400 to 1.4470
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in most common organic solvents
Formula Weight 130.14
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Ethyl 1-hydroxycyclopropanecarboxylate, 90%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.