missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-(hydroxymethyl)cyclopentanecarboxylate, 97%, Thermo Scientific Chemicals

Product Code. 15487856
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487856 1 g 1g
15497856 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487856 Supplier Thermo Scientific Alfa Aesar Supplier No. H62804.03
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

Ethyl 1-(hydroxymethyl)cyclopentanecarboxylate is employed as a intermediate for chemical research and synthetic building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 1-(hydroxymethyl)cyclopentanecarboxylate is employed as a intermediate for chemical research and synthetic building block.

Notes
Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1075-82-7
Molecular Formula C9H16O3
Molecular Weight (g/mol) 172.224
MDL Number MFCD12022629
InChI Key AUUICUZWYQYTNO-UHFFFAOYSA-N
Synonym ethyl 1-hydroxymethyl cyclopentanecarboxylate, ethyl 1-hydroxymethyl cyclopentane-1-carboxylate, 1-hydroxymethyl-cyclopentanecarboxylic acid ethyl ester
PubChem CID 46172884
IUPAC Name ethyl 1-(hydroxymethyl)cyclopentane-1-carboxylate
SMILES CCOC(=O)C1(CCCC1)CO

Specifications

Quantity 1 g
Formula Weight 172.22
Percent Purity 97%
Chemical Name or Material Ethyl 1-(hydroxymethyl)cyclopentanecarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.