missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-methyl-5-thien-2-yl-1H-pyrazole-3-carboxylate, 97%, Thermo Scientific™

Product Code. 10279282
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10279282 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10279282 Supplier Thermo Scientific Maybridge Supplier No. CC47123DA
 Restricted Item
Estimated Shipment: 03-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 868755-60-6
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H12N2O2S
MDL Number MFCD08060536
Quantity 1 g
Synonym ethyl 1-methyl-5-thien-2-yl-1h-pyrazole-3-carboxylate, ethyl 1-methyl-5-thiophen-2-yl pyrazole-3-carboxylate, ethyl 1-methyl-5-2-thienyl pyrazole-3-carboxylate, ethyl 1-methyl-5-thiophen-2-yl-1h-pyrazole-3-carboxylate, 1h-pyrazole-3-carboxylicacid,1-methyl-5-2-thienyl-,ethyl ester, 1h-pyrazole-3-carboxylicacid, 1-methyl-5-2-thienyl-, ethyl ester
InChI Key KIYMTNJEKXVPSQ-UHFFFAOYSA-N
SMILES CCOC(=O)C1=NN(C)C(=C1)C1=CC=CS1
IUPAC Name ethyl 1-methyl-5-thiophen-2-ylpyrazole-3-carboxylate
Molecular Weight (g/mol) 236.29
PubChem CID 7537650
Formula Weight 236.29
Percent Purity 97%
Chemical Name or Material Ethyl 1-methyl-5-thien-2-yl-1H-pyrazole-3-carboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.