missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,2-difluoropropionate, 97%

Produktkod. 15414507
Change view
Klicka för att se tillgängliga alternativ
Quantity:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Förpackningsstorlek
15414507 5 g 5g
15404507 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15414507 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H31852.06
 Restricted Item
Estimated Shipment: 02-12-2026
Lägg i varukorgen
Lägg i varukorgen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

CAS: 28781-85-3 | C5H8F2O2 | 138.114 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool dry place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 28781-85-3
Molecular Formula C5H8F2O2
Molecular Weight (g/mol) 138.114
MDL Number MFCD06657970
InChI Key ISVLDAIKRGXNCZ-UHFFFAOYSA-N
Synonym 2,2-difluoropropionic acid ethyl ester, ethyl 2,2-difluoropropionate, 2,2-difluoropropionicacidethylester, 2,2-difluoro-propionic acid ethyl ester, propanoic acid, 2,2-difluoro-, ethyl ester, ethyl difluoropropionate, ethyl 2,2 difluoropropionate, difluoropropionic acid ethyl ester, 2,2-difluoropropionic acidethyl ester
PubChem CID 2761188
IUPAC Name ethyl 2,2-difluoropropanoate
SMILES CCOC(=O)C(C)(F)F

Specifikationer

Density 1.102
Boiling Point 24°C (20 mmHg)
Linear Formula CH3CF2CO2CH2CH3
Quantity 5 g
UN Number UN3272
Solubility Information Insoluble in water.
Formula Weight 138.11
Percent Purity 97%
Chemical Name or Material Ethyl 2,2-difluoropropionate

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.