missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(2-methyl-1,3-dioxolan-2-yl)acetate, 98%, BLD Pharm™

Product Code. 31430961
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31430961 25 g
31424452 1 kg
31426381 500 g
31427458 100 g
31430217 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31430961 Supplier BLD Pharm Supplier No. BD10568225g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 6413-10-1
Molecular Formula C8H14O4
MDL Number MFCD00152488
Quantity 25 g
SMILES O=C(OCC)CC1(C)OCCO1
Molecular Weight (g/mol) 174.19
Percent Purity 98%
Chemical Name or Material Ethyl 2-(2-methyl-1,3-dioxolan-2-yl)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.