missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,4-dimethylbenzoate, 98+%, Thermo Scientific™

Product Code. 11327835
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11327835 5 g 5g
11337835 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 11327835

Brand: Thermo Scientific Alfa Aesar L02759.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33499-42-2
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00015438
InChI Key RJYPOWRKMKNFHH-UHFFFAOYSA-N
Synonym 2,4-dimethylbenzoic acid ethyl ester, benzoic acid, 2,4-dimethyl-, ethyl ester, acmc-20al1j, 2,4-dimethyl-benzoic acid ethyl ester, benzoic acid,2,4-dimethyl-, ethyl ester
PubChem CID 118492
IUPAC Name ethyl 2,4-dimethylbenzoate
SMILES CCOC(=O)C1=C(C=C(C=C1)C)C

Specifications

Quantity 5 g
Refractive Index 1.513
Beilstein 1938737
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 178.23
Percent Purity ≥98%
Chemical Name or Material Ethyl 2,4-dimethylbenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.