missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,4-dioxo-5-methylhexanoate, 95%

Product Code. 11471110
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471110 5 g 5g
11461110 1 g 1g
11481110 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11471110 Supplier Thermo Scientific Alfa Aesar Supplier No. B25735.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 64195-85-3 | C9H14O4 | 186.21 g/mol

Ethyl 2,4-dioxo-5-methylhexanoate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2,4-dioxo-5-methylhexanoate is used as a pharmaceutical intermediate.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64195-85-3
Molecular Formula C9H14O4
Molecular Weight (g/mol) 186.21
MDL Number MFCD07323638
InChI Key HBJPUWYQVWIKSD-UHFFFAOYSA-N
Synonym ethyl5-methyl-2,4-dioxohexanoate, ethyl 2,4-dioxo-5-methylhexanoate, 5-methyl-2,4-dioxohexanoic acid ethyl ester, hexanoic acid,5-methyl-2,4-dioxo-, ethyl ester, 5-methyl-2,4-dioxo-hexanoic acid ethyl ester, acmc-1bc7v, ethyl-2,4-dioxo-5-methylhexanoate, ethyl 5-methyl-2,4-dioxo-hexanoate, ethyl 5-methyl-2,4-bis oxidanylidene hexanoate
PubChem CID 3017501
IUPAC Name ethyl 5-methyl-2,4-dioxohexanoate
SMILES CCOC(=O)C(=O)CC(=O)C(C)C

Specifications

Quantity 5 g
Formula Weight 186.21
Percent Purity 95%
Chemical Name or Material Ethyl 2,4-dioxo-5-methylhexanoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.