missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(4-methylperhydro-1,4-diazepin-1-yl)benzoate, 97%, Thermo Scientific™

Product Code. 10545954
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10545954

Brand: Thermo Scientific Maybridge CC60423CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 912569-58-5
Molecular Formula C15H22N2O2
Molecular Weight (g/mol) 262.353
MDL Number MFCD09025904
InChI Key XAVHDPRFJKPWNA-UHFFFAOYSA-N
Synonym ethyl 2-4-methylperhydro-1,4-diazepin-1-yl benzoate, ethyl 2-4-methyl-1,4-diazepan-1-yl benzoate, ethyl 2-4-methyl-1,4-diazaperhydroepinyl benzoate
PubChem CID 18525910
IUPAC Name ethyl 2-(4-methyl-1,4-diazepan-1-yl)benzoate
SMILES CCOC(=O)C1=CC=CC=C1N2CCCN(CC2)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 262.35
Percent Purity 97%
Chemical Name or Material Ethyl 2-(4-methylperhydro-1, 4-diazepin-1-yl)benzoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.