missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-acetylisonicotinate, 96%

Product Code. 15447717
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15447717 5 g 5g
15437717 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15447717 Supplier Thermo Scientific Alfa Aesar Supplier No. H36911.06
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 25028-32-4 | C10H11NO3 | 193.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25028-32-4
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.20
MDL Number MFCD11042531
InChI Key LUMZQFHOBXCXOR-UHFFFAOYSA-N
Synonym ethyl 2-acetylisonicotinate, 2-acetyl-4-pyridinecarboxylic acid ethyl ester, pyac32, 2-acetyl-isonicotinic acid ethyl ester, 2-acetylpyridine-4-carboxylic acid ethyl ester
PubChem CID 55265486
IUPAC Name ethyl 2-acetylpyridine-4-carboxylate
SMILES CCOC(=O)C1=CC(=NC=C1)C(C)=O

Specifications

Melting Point 52°C to 55°C
Color Cream
Quantity 5 g
Formula Weight 193.2
Percent Purity 96%
Physical Form Fused Solid
Chemical Name or Material Ethyl 2-acetylisonicotinate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.