missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Amino-1,3-thiazole-4-carboxylate, TRC

Product Code. 30379159
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30379159 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30379159 Supplier Toronto Research Chemicals Supplier No. TRCE6784651G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 2-Amino-1,3-thiazole-4-carboxylate
CAS 5398-36-7
Molecular Formula C6H8N2O2S
Quantity 1 g
Synonym 2-Amino-1,3-thiazole-4-carboxylic Acid Ethyl Ester, 2-Amino-4-(ethoxycarbonyl)-1,3-thiazole, 2-Amino-4-ethoxycarbonylthiazole, 2-Amino-4-thiazolecarboxylic Acid Ethyl Ester, 2-Aminothiazol-4-carboxylic Acid Ethyl Ester, Ethyl 2-Amino-1,3-thiazole-4-carboxylate Ethyl 2-amino-4-thiazolecarboxylate, NSC 43547, NSC 4464
InChI Formula InChI=1S/C6H8N2O2S/c1-2-10-5(9)4-3-11-6(7)8-4/h3H,2H2,1H3,(H2,7,8)
SMILES CCOC(=O)c1csc(N)n1
Molecular Weight (g/mol) 172.2
Formula Weight 172.0306
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.