missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-1-cyclohexene-1-carboxylate, 98%, Thermo Scientific Chemicals

Product Code. 10757202 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10757202 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10757202 Supplier Thermo Scientific Acros Supplier No. 334260010
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1128-00-3 | Molecular Formula: C9H15NO2 | Molecular Weight: 169.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1128-00-3
Molecular Formula C9H15NO2
Molecular Weight (g/mol) 169.22
MDL Number MFCD01863243
InChI Key JBZVWABPSHNPIK-UHFFFAOYSA-N
Synonym ethyl 2-amino-1-cyclohexene-1-carboxylate, ethyl 2-aminocyclohex-1-enecarboxylate, 2-amino-cyclohex-1-enecarboxylic acid ethyl ester, ethyl 2-aminocyclohex-1-ene-1-carboxylate, 2-amino-1-cyclohexenecarboxylic acid ethyl ester, 1-cyclohexene-1-carboxylic acid, 2-amino-, ethyl ester, 2-aminocyclohex-1-enecarboxylic acid ethyl ester, 1-cyclohexene-1-carboxylicacid, 2-amino-, ethyl ester, 2-amino-cyclohex-1-enecarboxylicacidethylester, ethyl cis-2-aminocyclohexenecarboxylate
PubChem CID 312929
SMILES CCOC(=O)C1=C(N)CCCC1

Specifications

Melting Point 70°C to 74°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula NH2C6H8COOCH2CH3
Quantity 1 g
IUPAC Name ethyl 2-aminocyclohexene-1-carboxylate
Formula Weight 169.22
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Ethyl 2-amino-1-cyclohexene-1-carboxylate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.