missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-1-cyclopentene-1-carboxylate, 99%, Thermo Scientific™

Product Code. 10333063 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
500mg; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10333063 500mg; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10333063 Supplier Thermo Scientific Acros Supplier No. 292775000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7149-18-0
Melting Point 57°C to 60°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H13NO2
MDL Number MFCD00216929
Quantity 500 mg
Synonym ethyl 2-amino-1-cyclopentene-1-carboxylate, ethyl 2-aminocyclopent-1-enecarboxylate, 2-amino-cyclopent-1-enecarboxylic acid ethyl ester, ethyl 2-aminocyclopent-1-ene-1-carboxylate, 2-amino-cyclopent-1-enecarboxylicacidethylester, ethyl trans-2-aminocyclopentenecarboxylate, ethyl 2-amino-1-cyclopentene-1-carboxylate #, 2-aminocyclopentene-1-carboxylic acid ethyl ester, cyclopentene-1-carboxylic acid, 2-amino-, ethyl ester, 1-cyclopentene-1-carboxylicacid, 2-amino-, ethyl ester
Solubility Information Solubility in water: soluble. Other solubilities: soluble in chloroform,methanol and ethanol
InChI Key FMEHILPGLKGSCA-UHFFFAOYSA-N
SMILES CCOC(=O)C1=C(N)CCC1
IUPAC Name ethyl 2-aminocyclopentene-1-carboxylate
Molecular Weight (g/mol) 155.20
PubChem CID 243333
Formula Weight 155.2
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-amino-1-cyclopentene-1-carboxylate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 2-amino-1-cyclopentene-1-carboxylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.