missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-4-phenylthiophene-3-carboxylate, 96%

Product Code. 15424527
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15424527 1 g 1g
15417925 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15424527 Supplier Thermo Scientific Alfa Aesar Supplier No. H31867.03
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4815-36-5 | C13H13NO2S | 247.312 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4815-36-5
Molecular Formula C13H13NO2S
Molecular Weight (g/mol) 247.312
MDL Number MFCD00126391
InChI Key WYTHTMKMOSPACP-UHFFFAOYSA-N
Synonym 2-amino-4-phenyl-thiophene-3-carboxylic acid ethyl ester, 2-amino-4-phenylthiophene-3-carboxylic acid ethyl ester, ethyl 2-amino-4-phenyl-3-thiophenecarboxylate, thiophene-3-carboxylic acid, 2-amino-4-phenyl-, ethyl ester, amino-2 ethoxycarbonyl-3 phenyl-4 thiophene french, 3-thiophenecarboxylic acid, 2-amino-4-phenyl-, ethyl ester, ethyl 2-amino-4-phenyl-thiophene-3-carboxylate, 2-amino-4-phenyl-3-thiophenecarboxylic acid ethyl ester, cbmicro_024541, enamine_004212
PubChem CID 73242
IUPAC Name ethyl 2-amino-4-phenylthiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC=C1C2=CC=CC=C2)N

Specifications

Melting Point 90°C to 94°C
Quantity 1 g
Formula Weight 247.32
Percent Purity 96%
Chemical Name or Material Ethyl 2-amino-4-phenylthiophene-3-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.