missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate, 96%

Product Code. 15448785
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448785 1 g 1g
15458785 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448785 Supplier Thermo Scientific Alfa Aesar Supplier No. H34116.03
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 40106-13-6 | C12H17NO2S | 239.333 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in methanol and ethanol.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40106-13-6
Molecular Formula C12H17NO2S
Molecular Weight (g/mol) 239.333
MDL Number MFCD00216932
InChI Key XTUHIGALMIGZST-UHFFFAOYSA-N
Synonym ethyl 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta b thiophene-3-carboxylate, ethyl 2-amino-4h,5h,6h,7h,8h-cyclohepta b thiophene-3-carboxylate, ethyl 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta-b thiophene-3-carboxylate, ethyl 2-amino-4,5,6,7,8-pentahydrocyclohepta 1,2-b thiophene-3-carboxylate, 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta b thiophene-3-carboxylic acid ethyl ester, ethyl 2-aminocyclohepta b thiophene-3-carboxylate, aronis24232, ethyl 2-aminocycloheptathiophene-3-carboxylate, 2-amino-3-ethoxy-carbonyl-4,5-pentamethylenethiophene
PubChem CID 292644
IUPAC Name ethyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC2=C1CCCCC2)N

Specifications

Melting Point 87°C to 88°C
Quantity 1 g
Solubility Information Soluble in methanol and ethanol.
Formula Weight 239.33
Percent Purity 96%
Chemical Name or Material Ethyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.