missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-aminothiazole-5-carboxylate, 97%

Product Code. 15464397
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15464397 5 g 5g
15454397 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15464397 Supplier Thermo Scientific Alfa Aesar Supplier No. H31769.06
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 32955-21-8 | C6H8N2O2S | 172.202 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32955-21-8
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
MDL Number MFCD00602139
InChI Key VNZXERIGKZNEKB-UHFFFAOYSA-N
Synonym ethyl 2-aminothiazole-5-carboxylate, ethyl 2-amino-thiazole-5-carboxylate, 2-amino-thiazole-5-carboxylic acid ethyl ester, ethyl2-aminothiazole-5-carboxylate, 2-amino-thiazole-5-carboxylate ethyl ester, 2-aminothiazole-5-carboxylic acid ethyl ester, 5-thiazolecarboxylic acid, 2-amino-, ethyl ester, 2-amino-5-thiazolecarboxylic acid ethyl ester, ethyl-2-amino thiazole-5-carboxylate, zlchem 456
PubChem CID 314628
IUPAC Name ethyl 2-amino-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=CN=C(S1)N

Specifications

Melting Point 158°C to 160°C
Quantity 5 g
Sensitivity Light and moisture sensitive
Formula Weight 172.21
Percent Purity 97%
Chemical Name or Material Ethyl 2-aminothiazole-5-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.