missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-aminothiophene-3-carboxylate, 97%

Product Code. 15438128
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15438128 100 g 100g
15428128 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15438128 Supplier Thermo Scientific Alfa Aesar Supplier No. H61559.22
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 31891-06-2 | C7H9NO2S | 171.214 g/mol

Ethyl 2-aminothiophene-3-carboxylate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-aminothiophene-3-carboxylate is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31891-06-2
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.214
MDL Number MFCD01566303
InChI Key MKJQYFVTEPGXIE-UHFFFAOYSA-N
Synonym 2-amino-thiophene-3-carboxylic acid ethyl ester, 2-aminothiophene-3-carboxylic acid ethyl ester, ethyl2-aminothiophene-3-carboxylate, 2-amino-3-ethoxycarbonylthiophene, ethyl 2-amino-3-thiophenecarboxylate, 2-aminothiophene-3-carboxylate, 3-thiophenecarboxylic acid, 2-amino-, ethyl ester, pubchem22914, acmc-209hpg, ethyl 2-amino-3-thenoate
PubChem CID 1988156
IUPAC Name ethyl 2-aminothiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC=C1)N

Specifications

Quantity 100 g
Solubility Information Slightly soluble in water.
Formula Weight 171.22
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Ethyl 2-aminothiophene-3-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.