missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromothiazole-5-carboxylate, 98%

Product Code. 15451258
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15451258 5 g 5g
15461258 250 mg 250mg
15441258 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15451258 Supplier Thermo Scientific Alfa Aesar Supplier No. H55109.06
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 41731-83-3 | C6H6BrNO2S | 236.08 g/mol

Ethyl 2-bromothiazole-5-carboxylate is used in medicine, pharmaceutical intermediate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-bromothiazole-5-carboxylate is used in medicine, pharmaceutical intermediate

Solubility
Miscible is water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41731-83-3
Molecular Formula C6H6BrNO2S
Molecular Weight (g/mol) 236.08
MDL Number MFCD00463837
InChI Key KTYIFXLNIMPSKI-UHFFFAOYSA-N
PubChem CID 3614103
IUPAC Name ethyl 2-bromo-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=CN=C(Br)S1

Specifications

Density 1.581
Boiling Point 216°C to 217°C
Flash Point 110°C (230°F)
Refractive Index 1.553
Quantity 5 g
Synonym ethyl 2-bromothiazole-5-carboxylate, 2-bromo-thiazole-5-carboxylic acid ethyl ester, ethyl 2-bromo-5-thiazolecarboxylate, 2-bromothiazole-5-carboxylic acid ethyl ester, 2-bromo-5-ethoxycarbonyl thiazole, ethyl-2-bromothiazole-5-carboxylate, ethyl2-bromo-5-thiazolecarboxylate, ethyl2-bromothiazole-5-carboxylate, pubchem13480, acmc-1ambg
Solubility Information Miscible is water.
Formula Weight 236.08
Percent Purity 98%
Chemical Name or Material Ethyl 2-bromothiazole-5-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.